Learn More
Medchemexpress LLC HY-U00213 5mg Medchemexpress, Furaprofen CAS:67700-30-5 Purity:>98%
SDP
Catalog No. 502083192
Encompass_Preferred
Medchemexpress, HY-U00213 5mg Furaprofen CAS:67700-30-5 Furaprofen (R803) is an effective HCV replication inhibitor. Furaprofen (R803) is substantially more potent against genotype 1a and 1b replicons (EC50, ~30 nM) than against the genotype 2a replicon (EC50, ~1,000 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3192 Supplier Medchemexpress LLC Supplier No. HYU002135MG
Medchemexpress, HY-U00213 5mg Furaprofen CAS:67700-30-5 Furaprofen (R803) is an effective HCV replication inhibitor. Furaprofen (R803) is substantially more potent against genotype 1a and 1b replicons (EC50, ~30 nM) than against the genotype 2a replicon (EC50, ~1,000 nM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.