Learn More
FUJIFILM Biosciences Furathiocarb Standard
Catalog No. 5000450388
Encompass_Preferred
Furathiocarb Standard is a high-purity reference material intended for the accurate quantification of pesticide residues in food water and environmental samples This standard is ideal for gas chromatography GC and quantitative NMR qNMR applications requiring precise and reliable measurements Supplied as a solid with 99 0% purity GC and 98 0% by qNMR it is suited for use in regulatory compliance method validation and quality control protocols in pesticide monitoring The product is packaged under inert gas and should be stored between 2 10C protected from light Due to its acute toxicity and environmental hazards proper handling and disposal procedures must be followed
Catalog No. 50-004-50388 Supplier FUJIFILM Biosciences Supplier No. 06203981
Furathiocarb Standard is a high-purity reference material intended for the accurate quantification of pesticide residues in food water and environmental samples This standard is ideal for gas chromatography GC and quantitative NMR qNMR applications requiring precise and reliable measurements Supplied as a solid with 99 0% purity GC and 98 0% by qNMR it is suited for use in regulatory compliance method validation and quality control protocols in pesticide monitoring The product is packaged under inert gas and should be stored between 2 10C protected from light Due to its acute toxicity and environmental hazards proper handling and disposal procedures must be followed