Learn More
Cayman Chemical Furazolidone, Purity: ≥98%, CAS: 67-45-8, 50 g
SDP
Catalog No. 502948940
Encompass_Preferred
A nitrofuran antiprotozoal and antibacterial agent; active against G. lamblia trophozoites (IC50 = 2.9 µM), as well as metronidazole-susceptible and -resistant T. vaginalis clinical isolates (MLCs = Salmonella, Shigella, E. coli, and V. parahaemolyticus (MICs = ≤0.5-4 µg/ml); reduces splenic and hepatic parasite burden in a golden hamster model of L. chagasi infection
Catalog No. 50-294-8940 Supplier Cayman Chemical Supplier No. 3409050 G
A nitrofuran antiprotozoal and antibacterial agent; active against G. lamblia trophozoites (IC50 = 2.9 µM), as well as metronidazole-susceptible and -resistant T. vaginalis clinical isolates (MLCs = Salmonella, Shigella, E. coli, and V. parahaemolyticus (MICs = ≤0.5-4 µg/ml); reduces splenic and hepatic parasite burden in a golden hamster model of L. chagasi infection