Learn More
Selleck Chemical LLC Furazolidone S4528-100mg
SDP
Catalog No. 503111022
Encompass_Preferred
Furazolidone(Furoxone) is a synthetic nitrofuran derivative with bactericidal or bacteriostatic activity when used against Gram-positive and Gram-negative bacteria Furazolidone can be used to induce animal models of Cardiomyopathy
Catalog No. 50-311-1022 Supplier Selleck Chemical LLC Supplier No. S4528100MG
Furazolidone(Furoxone) is a synthetic nitrofuran derivative with bactericidal or bacteriostatic activity when used against Gram-positive and Gram-negative bacteria Furazolidone can be used to induce animal models of Cardiomyopathy