Learn More
Medchemexpress LLC Furazolidone | 500 MG
SDP
Catalog No. 5000412517
Encompass_Preferred
Furazolidone is a monoamine oxidase (MAO) inhibitor with antiproliferative, apoptosis-inducing and differentiation-promoting activities. It may inhibit leukemia fusion protein-mediated bone marrow transformation by upregulating the stability of the tumor suppressor protein p53, and exhibits anti-leukemic activity in acute myeloid leukemia (AML) cell lines.
  • Inhibits monoamine oxidase (MAO).
  • Demonstrates antiproliferative activity.
  • Induces apoptosis.
  • Promotes differentiation.
  • May inhibit leukemia fusion protein-mediated bone marrow transformation.
  • Upregulates stability of tumor suppressor protein p53.
  • Exhibits anti-leukemic activity in acute myeloid leukemia (AML) cell lines.
  • Suitable for anti-AML research.
Catalog No. 50-004-12517 Supplier Medchemexpress LLC Supplier No. HYB1336500MG
Furazolidone is a monoamine oxidase (MAO) inhibitor with antiproliferative, apoptosis-inducing and differentiation-promoting activities. It may inhibit leukemia fusion protein-mediated bone marrow transformation by upregulating the stability of the tumor suppressor protein p53, and exhibits anti-leukemic activity in acute myeloid leukemia (AML) cell lines.
  • Inhibits monoamine oxidase (MAO).
  • Demonstrates antiproliferative activity.
  • Induces apoptosis.
  • Promotes differentiation.
  • May inhibit leukemia fusion protein-mediated bone marrow transformation.
  • Upregulates stability of tumor suppressor protein p53.
  • Exhibits anti-leukemic activity in acute myeloid leukemia (AML) cell lines.
  • Suitable for anti-AML research.