Learn More
Medchemexpress LLC Furazolidone | 67-45-8 | 100.0% | 100 MG
SDP
Catalog No. 5000413241
Encompass_Preferred
Furazolidone is an analytical standard intended for research and analytical applications. This nitrofuran derivative exhibits antiprotozoal and antibacterial activity, and it inhibits AML1-ETO transformed cells with an IC50 of 12.7 μM.
  • Used as a reference standard in assays.
  • Utilized in qualitative, quantitative, and methodological research experiments.
  • Suitable for use with HPLC, GC, and MS.
Catalog No. 50-004-13241 Supplier Medchemexpress LLC Supplier No. HYB1336R100MG
Furazolidone is an analytical standard intended for research and analytical applications. This nitrofuran derivative exhibits antiprotozoal and antibacterial activity, and it inhibits AML1-ETO transformed cells with an IC50 of 12.7 μM.
  • Used as a reference standard in assays.
  • Utilized in qualitative, quantitative, and methodological research experiments.
  • Suitable for use with HPLC, GC, and MS.