Learn More
Medchemexpress LLC Furazolidone | 67-45-8 | MFCD00010550 | 1 ML
SDP
Catalog No. 5000413240
Encompass_Preferred
Furazolidone is a monoamine oxidase (MAO) inhibitor that possesses antiproliferative, apoptosis-inducing, and differentiation-promoting characteristics. It has the potential to inhibit leukemia fusion protein-mediated bone marrow transformation by increasing the stability of the tumor suppressor protein p53. Furthermore, Furazolidone demonstrates anti-leukemic activity in acute myeloid leukemia (AML) cell lines and is suitable for anti-AML research.
  • Monoamine oxidase (MAO) inhibitor.
  • Exhibits antiproliferative activity.
  • Induces apoptosis.
  • Promotes differentiation.
  • May inhibit leukemia fusion protein-mediated bone marrow transformation.
  • Upregulates the stability of tumor suppressor protein p53.
  • Shows anti-leukemic activity in acute myeloid leukemia (AML) cell lines.
  • Can be used for anti-AML research.
Catalog No. 50-004-13240 Supplier Medchemexpress LLC Supplier No. HYB133610MM/1ML
Furazolidone is a monoamine oxidase (MAO) inhibitor that possesses antiproliferative, apoptosis-inducing, and differentiation-promoting characteristics. It has the potential to inhibit leukemia fusion protein-mediated bone marrow transformation by increasing the stability of the tumor suppressor protein p53. Furthermore, Furazolidone demonstrates anti-leukemic activity in acute myeloid leukemia (AML) cell lines and is suitable for anti-AML research.
  • Monoamine oxidase (MAO) inhibitor.
  • Exhibits antiproliferative activity.
  • Induces apoptosis.
  • Promotes differentiation.
  • May inhibit leukemia fusion protein-mediated bone marrow transformation.
  • Upregulates the stability of tumor suppressor protein p53.
  • Shows anti-leukemic activity in acute myeloid leukemia (AML) cell lines.
  • Can be used for anti-AML research.