Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furazolidone, G-Biosciences
SCI_ED

Catalog No. S139729
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139729 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139729 Supplier G-Biosciences Supplier No. RC1028
Add to Cart
Add to Cart

Furazolidone is a nitrofuran derivative antibiotic with antiprotozoal and antibacterial activity. It targets a wide variety of bacteria, including those responsible for gastrointestinal infections.

It has also been used as a DNA cross-linking agent. Furazolidone binds bacterial DNA, inhibiting monoamine oxidase (MAO). It induces cross-links leading to an increased rate of mutation or halted DNA replication.

Chemical Identifiers

Molecular Weight (g/mol) 225.16 g/mol

Specifications

Boiling Point 400.48°C
Quantity 25 g
Percent Purity >98%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.