Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furazolidone, MP Biomedicals™

Catalog No. ICN19346525 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19346525 25 g
ICN19346501 1 g
ICN19346505 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19346525 Supplier MP Biomedicals, Inc Supplier No. 0219346525
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-45-8
Molecular Formula C8H7N3O5
Molecular Weight (g/mol) 225.16
InChI Key PLHJDBGFXBMTGZ-WEVVVXLNSA-N
Synonym furazolidone, furoxone, nitrofurazolidone, furazolidine, nitrofuroxon, furazolidon, diafuron, furaxone, furazol, furazon
PubChem CID 5323714
IUPAC Name 3-[(E)-(5-nitrofuran-2-yl)methylideneamino]-1,3-oxazolidin-2-one
SMILES C1COC(=O)N1N=CC2=CC=C(O2)[N+](=O)[O-]

Specifications

Color Yellow
Quantity 25 g
Formula Weight 225.2
Physical Form Powder
Chemical Name or Material Furazolidone
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.