Learn More
Medchemexpress LLC Furazolidone (Standard) | 67-45-8 | 99.95% | 50 MG
SDP
Catalog No. 5000413242
Encompass_Preferred
Furazolidone (Standard) is the analytical standard of Furazolidone, intended for research and analytical applications. It is a nitrofuran derivative with antiprotozoal and antibacterial activity, inhibiting AML1-ETO transformed cells with an IC50 of 12.7 μM.
  • Analytical standard of Furazolidone.
  • Intended for research and analytical applications.
  • Nitrofuran derivative.
  • Antiprotozoal and antibacterial activity.
  • Inhibits AML1-ETO transformed cells with an IC50 of 12.7 μM.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.
Catalog No. 50-004-13242 Supplier Medchemexpress LLC Supplier No. HYB1336R50MG
Furazolidone (Standard) is the analytical standard of Furazolidone, intended for research and analytical applications. It is a nitrofuran derivative with antiprotozoal and antibacterial activity, inhibiting AML1-ETO transformed cells with an IC50 of 12.7 μM.
  • Analytical standard of Furazolidone.
  • Intended for research and analytical applications.
  • Nitrofuran derivative.
  • Antiprotozoal and antibacterial activity.
  • Inhibits AML1-ETO transformed cells with an IC50 of 12.7 μM.
  • Commonly used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS.