Learn More
Medchemexpress LLC HY-106080A 10mg Medchemexpress, Furegrelate (sodium) CAS:85666-17-7 Purity:>98%
SDP
Catalog No. 501968315
Encompass_Preferred
Medchemexpress, HY-106080A 10mg Furegrelate (sodium) CAS:85666-17-7 Furegrelate Sodium (U-63557A) is a potent, orally available, and selective thromboxane synthase inhibitor. Furegrelate Sodium inhibits human platelet microsomal thromboxane A2 (TxA2) synthase with an IC50 of 15 nM. Furegrelate Sodium is being developed as an antiplatelet agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8315 Supplier Medchemexpress LLC Supplier No. HY106080A10MG
Medchemexpress, HY-106080A 10mg Furegrelate (sodium) CAS:85666-17-7 Furegrelate Sodium (U-63557A) is a potent, orally available, and selective thromboxane synthase inhibitor. Furegrelate Sodium inhibits human platelet microsomal thromboxane A2 (TxA2) synthase with an IC50 of 15 nM. Furegrelate Sodium is being developed as an antiplatelet agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.