Learn More
TARGETMOL CHEMICALS INC FUREGRELATE SODIUM 5MG
SDP
Catalog No. 502581541
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Furegrelate sodium (U-63557A) is a selective thromboxane synthase inhibitor with oral activity. Furegrelate sodium(U-63557A) inhibits thromboxane A2 (TXA2) synthase in human platelet microsomes with an IC50 of 15 nM. purity: 100%
Catalog No. 50-258-1541 Supplier TARGETMOL CHEMICALS INC Supplier No. T113395MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Furegrelate sodium (U-63557A) is a selective thromboxane synthase inhibitor with oral activity. Furegrelate sodium(U-63557A) inhibits thromboxane A2 (TXA2) synthase in human platelet microsomes with an IC50 of 15 nM. purity: 100%