Learn More
Medchemexpress LLC Furegrelate sodium | 85666-17-7 | MFCD00058448 | 99.8% | 275.23 | C15H10NNaO3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY106080A10MM 1ML
Encompass_Preferred
Furegrelate sodium is the sodium salt form of furegrelate, a potent and selective thromboxane synthase inhibitor used in research to inhibit thromboxane A2 production and modulate platelet aggregation. The compound is supplied for laboratory use in solution and solid formats and is characterized by a defined molecular formula and high purity.
  • Potent, selective thromboxane synthase inhibitor.
  • Sodium salt form for research applications.
  • Supplied as 10 mM solution in DMSO and as solid quantities.
  • High purity (99.8%).
  • Molecular weight 275.23 g/mol.
Catalog No. 50-002-42712