Learn More
Cayman Chemical Furegrelate (sodium salt), Purity: ≥99%, CAS: 85666-17-7, 50 mg
SDP
Catalog No. 502893524
Encompass_Preferred
Furegrelate is a potent inhibitor of thromboxane synthase with little effect on other enzymes essential for arachidonate metabolism. The IC50 value is 15 nM for human platelet microsomal thromboxane synthase.{1227,1228}
Catalog No. 50-289-3524 Supplier Cayman Chemical Supplier No. 7054050 MG
Furegrelate is a potent inhibitor of thromboxane synthase with little effect on other enzymes essential for arachidonate metabolism. The IC50 value is 15 nM for human platelet microsomal thromboxane synthase.{1227,1228}