Learn More
Medchemexpress LLC Furfuryl acetate | 623-17-6 | 98.3% | 140.14 | 100 G
SDP
Catalog No. 5000452535
Encompass_Preferred
Furfuryl acetate is a chemical compound primarily utilized in synthetic organic chemistry. It serves as a key intermediate in the creation of more complex furan derivatives.
  • Can be used in the synthesis of 5-acetoxymethyl-2-vinylfuran.
  • Can be used in the synthesis of 5-hydroxymethyl-2-vinylfuran.
  • Appears as a colorless to light yellow liquid.
  • Exhibits solubility in DMSO at concentrations of 100 mg/mL or greater.
  • Recommended storage conditions optimize stability for both pure form and in solvent.
Catalog No. 50-004-52535 Supplier Medchemexpress LLC Supplier No. HYW010321100G
Furfuryl acetate is a chemical compound primarily utilized in synthetic organic chemistry. It serves as a key intermediate in the creation of more complex furan derivatives.
  • Can be used in the synthesis of 5-acetoxymethyl-2-vinylfuran.
  • Can be used in the synthesis of 5-hydroxymethyl-2-vinylfuran.
  • Appears as a colorless to light yellow liquid.
  • Exhibits solubility in DMSO at concentrations of 100 mg/mL or greater.
  • Recommended storage conditions optimize stability for both pure form and in solvent.