Learn More
Medchemexpress LLC Furfuryl acetate | 623-17-6 | 98.3% | 140.14 | 25 G
SDP
Catalog No. 5000454124
Encompass_Preferred
Furfuryl acetate is a laboratory chemical used for the synthesis of substances such as 5-acetoxymethyl-2-vinylfuran and 5-hydroxymethyl-2-vinylfuran. It is intended for research use only and should be handled by qualified personnel in appropriate facilities, as it has not been fully validated for medical applications.
  • Used in the synthesis of 5-acetoxymethyl-2-vinylfuran.
  • Used in the synthesis of 5-hydroxymethyl-2-vinylfuran.
  • Intended for laboratory use.
  • Harmful if swallowed.
  • Causes skin irritation.
  • Causes serious eye irritation.
  • May cause respiratory irritation.
Catalog No. 50-004-54124 Supplier Medchemexpress LLC Supplier No. 25GHYW010321
Furfuryl acetate is a laboratory chemical used for the synthesis of substances such as 5-acetoxymethyl-2-vinylfuran and 5-hydroxymethyl-2-vinylfuran. It is intended for research use only and should be handled by qualified personnel in appropriate facilities, as it has not been fully validated for medical applications.
  • Used in the synthesis of 5-acetoxymethyl-2-vinylfuran.
  • Used in the synthesis of 5-hydroxymethyl-2-vinylfuran.
  • Intended for laboratory use.
  • Harmful if swallowed.
  • Causes skin irritation.
  • Causes serious eye irritation.
  • May cause respiratory irritation.