Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furfuryl Methacrylate (stabilized with MEHQ) 95.0+%, TCI America™
SDP

Catalog No. M133525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M133525G 25 g
M13355G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M133525G Supplier TCI America Supplier No. M133525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3454-28-2
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00046113
InChI Key DWXAVNJYFLGAEF-UHFFFAOYSA-N
Synonym Methacrylic Acid Furfuryl Ester
PubChem CID 76993
IUPAC Name furan-2-ylmethyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCC1=CC=CO1

Specifications

Color Yellow
Boiling Point 82°C
Quantity 25 g
Formula Weight 166.18
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Furfuryl Methacrylate (stabilized with MEHQ)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.