Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furfuryl methyl sulfide, 98%, Thermo Scientific™

Catalog No. AAB2169122
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2169122 100 g
AAB2169106 5 g
AAB2169114 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2169122 Supplier Thermo Scientific Chemicals Supplier No. B2169122

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1438-91-1
Molecular Formula C6H8OS
Molecular Weight (g/mol) 128.19
MDL Number MFCD00009604
InChI Key SKSFHXVDHVKIBN-UHFFFAOYSA-N
Synonym furfuryl methyl sulfide, methyl furfuryl sulfide, 2-methylthio methyl furan, 2-furfuryl methyl sulfide, furan, 2-methylthio methyl, 2-methylsulfanylmethyl furan, unii-lmb8355w32, 2-methylsulfanyl methyl furan, 2-methylthio methylfuran
PubChem CID 518937
SMILES CSCC1=CC=CO1

Specifications

Density 1.07
Boiling Point 170°C to 173°C
Flash Point 63°C (145°F)
Refractive Index 1.524
Quantity 100 g
Beilstein 107109
IUPAC Name 2-(methylsulfanylmethyl)furan
Formula Weight 128.19
Percent Purity 98%
Chemical Name or Material Furfuryl methyl sulfide
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 215-874-0
TSCA Yes
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.