Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furfuryl Methyl Sulfide 98.0+%, TCI America™
SDP

Catalog No. F024525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
F024525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. F024525G Supplier TCI America Supplier No. F024525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1438-91-1
Molecular Formula C6H8OS
Molecular Weight (g/mol) 128.19
MDL Number MFCD00009604
InChI Key SKSFHXVDHVKIBN-UHFFFAOYSA-N
Synonym furfuryl methyl sulfide, methyl furfuryl sulfide, 2-methylthio methyl furan, 2-furfuryl methyl sulfide, furan, 2-methylthio methyl, 2-methylsulfanylmethyl furan, unii-lmb8355w32, 2-methylsulfanyl methyl furan, 2-methylthio methylfuran
PubChem CID 518937
IUPAC Name 2-[(methylsulfanyl)methyl]furan
SMILES CSCC1=CC=CO1

Specifications

Color Yellow
Boiling Point 65°C
Quantity 25 g
Formula Weight 128.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Furfuryl Methyl Sulfide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.