Learn More
AdipoGen Furil (5 g)
SDP
Supplier:  AdipoGen CDXF0114G005
Encompass_Preferred
Chemical. CAS: 492-94-4. Formula: C10H6O4. MW: 190.15. Heterocyclic building block for synthesis. Furil undergoes catalytic asymmetric transfer hydrogenation to yield hydrofuroin.
Catalog No. 50-205-4441