Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ FURIN (Human) ELISA Kit

Catalog No. 89101040 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
96 Assays
Sample Type:
CCS, Cell Lysates, Tissue Lysates
CCS, Plasma, Serum, Urine
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Sample Type
89-101-040 1 Kit CCS, Cell Lysates, Tissue Lysates
89-028-570 96 Assays CCS, Plasma, Serum, Urine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-101-040 Supplier Abnova™ Supplier No. KA3361
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

In-Vitro Enzyme-Linked Iimmunosorbent assay for the quantitative measurement of Human FURIN.

Specifications

Calibration Range 312 to 20000pg/mL
Cross Reactivity Human
Description FURIN (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human FURIN.
Detection Method Colorimetric
Quantity 1 Kit
Sample Type CCS, Cell Lysates, Tissue Lysates
Sample Volume 100 μL
Content And Storage Store the kit at -20°C
For Use With (Application) Quantification
Product Type FURIN (Human) ELISA Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.