Learn More
Apexbio Technology LLC Furosemide,1g CAS# 54-31-9
SDP
Supplier:  Apexbio Technology LLC A84351000
Encompass_Preferred
Furosemide is an inhibitor of Na+/2Cl-/K+ cotransporter [1].The Na-K-Cl cotransporter (NKCC) is a protein that acts as an active transporter helping sodium, potassium, and chloride into and out of cells. NKCC1 has important functions in organs that secrete fluids. NKCC2 exists in the kidney and serves to extract sodium, potassium, and chloride from the urine [2].Furosemide is an antagonist of Na+/2Cl-/K+ cotransporter. In adult brain tissue, furosemide blocked the epileptiform activity through inhibiting cell swelling, which was concomitant with its inhibition of activity-driven changes of the extracellular space (ECS) [1]. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-224-0317