Learn More
U.S. Pharmacopeia Furosemide, 54-31-9, MFCD00010549, 200 mg
Supplier:  U.S. Pharmacopeia 1287008
Encompass
Empirical Formula (Hill Notation): C12H11ClN2O5S, Molecular Weight: 330.74, mp: 220 °C, Synonym: 4-Chloro-N-furfuryl-5-sulfamoylanthranilic acid, 5-(Aminosulfonyl)-4-chloro-2-([2-furanylmethyl]amino)benzoic acid.
Catalog No. NC1403957