Learn More
Medchemexpress LLC Furosemide | 54-31-9 | 99.8% | 1 ML
SDP
Catalog No. 5000450467
Encompass_Preferred
Furosemide is a potent and orally active inhibitor of Na+/K+/2Cl- (NKCC) cotransporter, specifically NKCC1 and NKCC2. It also acts as a GABAA receptor antagonist, showing 100-fold selectivity for α6-containing receptors compared to α1-containing receptors. Furosemide functions as a loop diuretic and is used for the study of congestive heart failure, hypertension, and edema.
  • Potent and orally active inhibitor of NKCC1 and NKCC2 cotransporters.
  • GABAA receptors antagonist.
  • Displays 100-fold selectivity for α6-containing GABAA receptors over α1-containing receptors.
  • Acts as a loop diuretic.
Catalog No. 50-004-50467 Supplier Medchemexpress LLC Supplier No. HYB013510MM/1ML
Furosemide is a potent and orally active inhibitor of Na+/K+/2Cl- (NKCC) cotransporter, specifically NKCC1 and NKCC2. It also acts as a GABAA receptor antagonist, showing 100-fold selectivity for α6-containing receptors compared to α1-containing receptors. Furosemide functions as a loop diuretic and is used for the study of congestive heart failure, hypertension, and edema.
  • Potent and orally active inhibitor of NKCC1 and NKCC2 cotransporters.
  • GABAA receptors antagonist.
  • Displays 100-fold selectivity for α6-containing GABAA receptors over α1-containing receptors.
  • Acts as a loop diuretic.