Learn More
Medchemexpress LLC Furosemide | 54-31-9 | 99.83% | 1 G
SDP
Catalog No. 5000450925
Encompass_Preferred
Furosemide is a potent and orally active inhibitor of Na+/K+/2Cl- (NKCC) cotransporter, specifically NKCC1 and NKCC2. It also acts as a GABAA receptors antagonist, displaying 100-fold selectivity for α6-containing receptors over α1-containing receptors. This compound functions as a loop diuretic and is widely used for the study of congestive heart failure, hypertension, and edema.
  • Potent and orally active inhibitor of NKCC cotransporter (NKCC1 and NKCC2)
  • GABAA receptors antagonist
  • Displays 100-fold selectivity for α6-containing receptors
  • Acts as a loop diuretic
  • Used for studies of congestive heart failure
  • Used for studies of hypertension
  • Used for studies of edema
Catalog No. 50-004-50925 Supplier Medchemexpress LLC Supplier No. HYB01351G
Furosemide is a potent and orally active inhibitor of Na+/K+/2Cl- (NKCC) cotransporter, specifically NKCC1 and NKCC2. It also acts as a GABAA receptors antagonist, displaying 100-fold selectivity for α6-containing receptors over α1-containing receptors. This compound functions as a loop diuretic and is widely used for the study of congestive heart failure, hypertension, and edema.
  • Potent and orally active inhibitor of NKCC cotransporter (NKCC1 and NKCC2)
  • GABAA receptors antagonist
  • Displays 100-fold selectivity for α6-containing receptors
  • Acts as a loop diuretic
  • Used for studies of congestive heart failure
  • Used for studies of hypertension
  • Used for studies of edema