Learn More
Medchemexpress LLC Furosemide-d5 | 1189482-35-6 | 99.9% | 1 MG
SDP
Catalog No. 5000450887
Encompass_Preferred
Furosemide-d5 is the deuterium labeled Furosemide. Furosemide is a potent and orally active inhibitor of Na+/K+/2Cl- (NKCC) cotransporter, specifically NKCC1 and NKCC2. It also acts as a GABAA receptors antagonist, showing 100-fold selectivity for α6-containing receptors over α1-containing receptors. Furosemide is a loop diuretic used in the study of congestive heart failure, hypertension, and edema. Deuterium substitution in drug molecules is explored for its potential impact on pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potent and orally active inhibitor of Na+/K+/2Cl- (NKCC) cotransporter
  • Acts as a GABAA receptors antagonist
  • Used in the study of congestive heart failure, hypertension, and edema
Catalog No. 50-004-50887 Supplier Medchemexpress LLC Supplier No. HYB0135S1MG
Furosemide-d5 is the deuterium labeled Furosemide. Furosemide is a potent and orally active inhibitor of Na+/K+/2Cl- (NKCC) cotransporter, specifically NKCC1 and NKCC2. It also acts as a GABAA receptors antagonist, showing 100-fold selectivity for α6-containing receptors over α1-containing receptors. Furosemide is a loop diuretic used in the study of congestive heart failure, hypertension, and edema. Deuterium substitution in drug molecules is explored for its potential impact on pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Potent and orally active inhibitor of Na+/K+/2Cl- (NKCC) cotransporter
  • Acts as a GABAA receptors antagonist
  • Used in the study of congestive heart failure, hypertension, and edema