Learn More
Selleck Chemical LLC Furosemide sodium S5194-25mg
SDP
Catalog No. 503112414
Encompass_Preferred
Furosemide (Frusemide) sodium is a potent and orally active inhibitor of Na-K-Cl cotransporter (NKCC) Furosemide sodium is a subtype-selective antagonist of gamma-aminobutyric acid type A (GABAA) receptor
Catalog No. 50-311-2414 Supplier Selleck Chemical LLC Supplier No. S519425MG
Furosemide (Frusemide) sodium is a potent and orally active inhibitor of Na-K-Cl cotransporter (NKCC) Furosemide sodium is a subtype-selective antagonist of gamma-aminobutyric acid type A (GABAA) receptor