Learn More
Medchemexpress LLC Furosemide (sodium) | 41733-55-5 | 99.9% | 1 G
SDP
Catalog No. 5000450856
Encompass_Preferred
Furosemide sodium is a potent and orally active inhibitor of Na+/K+/2Cl- (NKCC) cotransporter, specifically NKCC1 and NKCC2. It also acts as a GABAA receptor antagonist, showing 100-fold selectivity for α6-containing receptors over α1-containing receptors. This compound functions as a loop diuretic and is used in studies related to congestive heart failure, hypertension, and edema. It is intended for research use only and is not sold to patients.
  • Potent and orally active inhibitor of Na+/K+/2Cl- cotransporter
  • Acts as a GABAA receptor antagonist
  • Shows 100-fold selectivity for α6-containing receptors
  • Functions as a loop diuretic
  • Used in studies related to congestive heart failure, hypertension, and edema
Catalog No. 50-004-50856 Supplier Medchemexpress LLC Supplier No. HYB0135A1G
Furosemide sodium is a potent and orally active inhibitor of Na+/K+/2Cl- (NKCC) cotransporter, specifically NKCC1 and NKCC2. It also acts as a GABAA receptor antagonist, showing 100-fold selectivity for α6-containing receptors over α1-containing receptors. This compound functions as a loop diuretic and is used in studies related to congestive heart failure, hypertension, and edema. It is intended for research use only and is not sold to patients.
  • Potent and orally active inhibitor of Na+/K+/2Cl- cotransporter
  • Acts as a GABAA receptor antagonist
  • Shows 100-fold selectivity for α6-containing receptors
  • Functions as a loop diuretic
  • Used in studies related to congestive heart failure, hypertension, and edema