Learn More
Medchemexpress LLC Furosemide (Standard) | 54-31-9 | 99.9% | 25 MG
SDP
Catalog No. 5000406817
Encompass_Preferred
Furosemide (Standard) is an analytical standard of Furosemide with a purity of 99.95%. It is a potent and orally active inhibitor of Na+/K+/2Cl- (NKCC) cotransporter, specifically NKCC1 and NKCC2, and also acts as a GABAA receptor antagonist. This compound functions as a loop diuretic and is used in the study of congestive heart failure, hypertension, and edema, serving as a reference for qualitative, quantitative, and methodological research experiments.
  • Intended for research and analytical applications
  • Potent and orally active inhibitor of Na+/K+/2Cl- (NKCC) cotransporter
  • Acts as a GABAA receptor antagonist
  • Functions as a loop diuretic
  • Used in the study of congestive heart failure, hypertension, and edema
  • Analytical standard for qualitative, quantitative, and methodological research experiments
Catalog No. 50-004-06817 Supplier Medchemexpress LLC Supplier No. 25MGHYB0135R
Furosemide (Standard) is an analytical standard of Furosemide with a purity of 99.95%. It is a potent and orally active inhibitor of Na+/K+/2Cl- (NKCC) cotransporter, specifically NKCC1 and NKCC2, and also acts as a GABAA receptor antagonist. This compound functions as a loop diuretic and is used in the study of congestive heart failure, hypertension, and edema, serving as a reference for qualitative, quantitative, and methodological research experiments.
  • Intended for research and analytical applications
  • Potent and orally active inhibitor of Na+/K+/2Cl- (NKCC) cotransporter
  • Acts as a GABAA receptor antagonist
  • Functions as a loop diuretic
  • Used in the study of congestive heart failure, hypertension, and edema
  • Analytical standard for qualitative, quantitative, and methodological research experiments