Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Furosemide, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18602876 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-876 5 g Amber Glass Bottle
18-602-877 25 g Amber Glass Bottle
18-602-878 100 g Amber Glass Bottle
18-602-879 500 g Amber Glass Bottle
18-602-880 1 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-602-876 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F11335GM
Add to Cart
Add to Cart

F1133, 54-31-9, C12H11ClN2O5S

Spectrum™ Chemical Furosemide, USP is used to reduce extra fluid in the body (edema) caused by conditions such as heart failure, liver disease, and kidney disease. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 54-31-9
Molecular Formula C12H11ClN2O5S
Molecular Weight (g/mol) 330.74
InChI Key ZZUFCTLCJUWOSV-UHFFFAOYSA-N
IUPAC Name 4-chloro-2-{[(furan-2-yl)methyl]amino}-5-sulfamoylbenzoic acid
SMILES NS(=O)(=O)C1=CC(C(O)=O)=C(NCC2=CC=CO2)C=C1Cl

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 1%
Packaging Amber Glass Bottle
Quantity 5 g
Percent Purity 98 to 102%
Grade USP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.