Learn More
Enzo Life Sciences Furoxan (10mg). CAS: 125520-62-9
SDP
Catalog No. 502010462
Encompass_Preferred
A novel NO donor which only releases NO in the presence of thiols. It is a potent vasodilator and inhibitor of platelet aggregation. Alternative name: 4-Phenyl-3-furoxancarbonitrile. Purity: ≥99% (HPLC). Solubility: Soluble in ethanol (25 mg/ml) or DMSO (25 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0462 Supplier Enzo Life Sciences Supplier No. BMLCN2550010
A novel NO donor which only releases NO in the presence of thiols. It is a potent vasodilator and inhibitor of platelet aggregation. Alternative name: 4-Phenyl-3-furoxancarbonitrile. Purity: ≥99% (HPLC). Solubility: Soluble in ethanol (25 mg/ml) or DMSO (25 mg/ml). Long Term Storage: -20°C.