Learn More
Medchemexpress LLC HY-106907 10mg Medchemexpress, Furprofen CAS:66318-17-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10690710MG
Encompass_Preferred
Medchemexpress, HY-106907 10mg Furprofen CAS:66318-17-0 Furprofen is an non-steroidal anti-inflammatory drug (NSAID) with analgesic properties. Furprofen acts via the inhibition of prostaglandin (PGE) synthesis. Furprofen can be treated orally for the relief of pain[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8358