Learn More
Cayman Chemical Fursultiamine, Purity: ≥95%, CAS: 804-30-8, 100 mg
SDP
Catalog No. 502948247
Encompass_Preferred
A synthetic derivative of vitamin B1; reduces LPS-induced increases in IL-6, IL-8, and CCL2 at 100 and 20 µM, respectively; reduces the area and severity of laser-induced lesions in a mouse model of CNV at 50 mg/kg; prevents physical-fatigue loading decreases in skeletal muscle ATP levels induced by forced swimming with a weight load in rats at 50 mg/kg; enhances the protective effects of glucosamine and chondroitin sulfate on cartilage lesion formation in a rabbit model of osteoarthritis at 100 mg/kg
Catalog No. 50-294-8247 Supplier Cayman Chemical Supplier No. 33456100 MG
A synthetic derivative of vitamin B1; reduces LPS-induced increases in IL-6, IL-8, and CCL2 at 100 and 20 µM, respectively; reduces the area and severity of laser-induced lesions in a mouse model of CNV at 50 mg/kg; prevents physical-fatigue loading decreases in skeletal muscle ATP levels induced by forced swimming with a weight load in rats at 50 mg/kg; enhances the protective effects of glucosamine and chondroitin sulfate on cartilage lesion formation in a rabbit model of osteoarthritis at 100 mg/kg