Learn More
Aviva Systems Biology Corp FUS Antibody - N-terminal region (ARP40278_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40278_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against FUS. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human FUS. Alternate names: FUS-CHOP, FUS1, TLS, ALS6, POMP75, HNRNPP2ApplicationIHC, WB. Species reactivity: Cow, Dog, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-53186