Learn More
Montana Molecular FUS WT BacMam - 5mL
SDP
Supplier:  Montana Molecular N0300N005
Encompass_Preferred
Wild-type FUS packaged in BacMam for expression in mammalian cells. BacMam provides even, near 100% transduction efficiency in HEK293 cells as well as expression in many other cell lines, iPSCs, and primary cultures. Expression is easily titrateable to your experimental needs or to adjust assay sensitivity. A version with a P525L mutation is also available. Co-express with Montana Molecular sensors for ER stress, cAMP, calcium, and more. This kit includes the Wild-type FUS BacMam and 500mM sodium butyrate in H2O.
Catalog No. NC2600426