Learn More
Apexbio Technology LLC Fusaric Acid 536-69-6 500mg
SDP
Catalog No. 5000064083
Encompass_Preferred
Fusaric acid is a mycotoxin metabolite derived from several Fusarium species chemically characterized as a pyridine derivative Biochemically fusaric acid functions primarily as an inhibitor of dopamine -hydroxylase the enzyme responsible for converting dopamine into norepinephrine By inhibiting dopamine -hydroxylase activity fusaric acid causes reductions of endogenous norepinephrine and epinephrine in biological tissues including the brain adrenal glands heart and spleen In experimental research fusaric acid is utilized to study neurotransmitter metabolism catecholamine pathways and physiological effects associated with altered norepinephrine synthesis Additionally fusaric acid administration in animal models has been shown to affect serotonin and dopamine metabolism in neural tissues and may synergistically interact with other mycotoxins making it relevant in toxicological and pharmacological studies
Catalog No. 50-000-64083 Supplier Apexbio Technology LLC Supplier No. C5062500
Add to Cart
Add to Cart
Fusaric acid is a mycotoxin metabolite derived from several Fusarium species chemically characterized as a pyridine derivative Biochemically fusaric acid functions primarily as an inhibitor of dopamine -hydroxylase the enzyme responsible for converting dopamine into norepinephrine By inhibiting dopamine -hydroxylase activity fusaric acid causes reductions of endogenous norepinephrine and epinephrine in biological tissues including the brain adrenal glands heart and spleen In experimental research fusaric acid is utilized to study neurotransmitter metabolism catecholamine pathways and physiological effects associated with altered norepinephrine synthesis Additionally fusaric acid administration in animal models has been shown to affect serotonin and dopamine metabolism in neural tissues and may synergistically interact with other mycotoxins making it relevant in toxicological and pharmacological studies