Learn More
Cayman Chemical Enniatin A, Purity: ≥99%, CAS: 2503-13-1, 2.5 mg
SDP
Catalog No. 502965677
Encompass_Preferred
One of four major analogs in the enniatin complex that has been shown to moderately inhibit ACAT activity in rat liver microsomes (IC50 = 22 µM); demonstrates anthelmintic properties against N. brasiliensis, T. spiralis, and H. spumosa
Catalog No. 50-296-5677 Supplier Cayman Chemical Supplier No. 174562.5 MG
One of four major analogs in the enniatin complex that has been shown to moderately inhibit ACAT activity in rat liver microsomes (IC50 = 22 µM); demonstrates anthelmintic properties against N. brasiliensis, T. spiralis, and H. spumosa