Learn More
ATCC Lactobacillus leichmannii
Supplier:  ATCC 4797
Encompass
Lactobacillus leichmannii strain 326 is a bacterial type strain that metabolizes arginine. This strain is propagated in a 5% CO2 atmosphere in lactobacilli MRS media.
Catalog No. 50-237-9738