Learn More
ATCC Pfizer Pneumococcal polysaccharide powder Type 20 (d20) 2mg
Catalog No. 502379633
Encompass
Purified pneumococcal polysaccharide type 20 is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 20 can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-237-9633 Supplier ATCC Supplier No. 524X
Purified pneumococcal polysaccharide type 20 is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 20 can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.