Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZeptoMetrix Fusarium Solani Z081, Titered
SDP

Catalog No. 22032198 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-032-198 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-032-198 Supplier ZeptoMetrix Supplier No. 0801806
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Live, titered Fusarium solani for LOD determination, diagnostic assay development, and cross-reactivity studies.

  • Each frozen aliquot contains 1 mL of pure, titered Fusarium solani culture
  • Organism identity confirmed by ITS2 sequencing
  • Culture purity monitored through staining and additional culturing
  • Titer determined using a minimum of two aliquots after freezing
  • Freezing medium contains 15% glycerol as a cryoprotectant
  • Certificate of Analysis provides details on the specific freezing medium used
  • When used as a nucleic acid test control, the same amplification protocols used for clinical specimens should be followed

Specifications

Product Type Bacteria Control
Sterility Non-sterile
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.