Learn More
AdipoGen Fuscin (2.5 mg)
SDP
Supplier:  AdipoGen AGCN20138MM25
Encompass_Preferred
Chemical. CAS: 83-85-2. Formula: C15H16O5. MW: 276.3. Isolated from Oidiodendron sp. Mycotoxin. Antibacterial agent. ADP transport inhibitor. Mitochondrial respiration and oxidative phosphorylation inhibitor. Human CCR5 receptor antagonist. Anti-HIV compound.
Catalog No. 50-205-3514