Learn More
Perkin Elmer US LLC 0.53 mm, 5 m, Fused Silica Tubing Intermediate Polarity
Catalog No. 509045822
Encompass_Preferred
This tubing features non-polar deactivation for injection solvents such as alkanesand ethers. The intermediate polar deactivation is useful for injection solvents such as acetone, dichloromethane, and toluene. The polar deactivation for injection solvents like water. The diameter for this tubing is 0.53 mm and is 5 meters long.
Catalog No. 50-904-5822 Supplier Perkin Elmer US LLC Supplier No. N9301358
This tubing features non-polar deactivation for injection solvents such as alkanesand ethers. The intermediate polar deactivation is useful for injection solvents such as acetone, dichloromethane, and toluene. The polar deactivation for injection solvents like water. The diameter for this tubing is 0.53 mm and is 5 meters long.