Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Fuse, 10A, 5 × 20 mm, Slo-Blo

Catalog No. 50792769 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This fuse is a Waters Quality part designed for various applications requiring specific electrical protection. It features a 10A rating and dimensions of 5 × 20 mm, with a slo-blo characteristic for delayed circuit protection. It is provided as a spare part to ensure operational continuity.

  • Engineering code: 330000122
  • UNSPSC code: 39121600
  • Product type: Spare parts kits
  • Units per package: 1 pk
  • Replacement schedule: As required

Catalog No. 50-792-769 Supplier Waters Corp Supplier No. 700002577
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This fuse is a Waters Quality part designed for various applications requiring specific electrical protection. It features a 10A rating and dimensions of 5 × 20 mm, with a slo-blo characteristic for delayed circuit protection. It is provided as a spare part to ensure operational continuity.

  • Engineering code: 330000122
  • UNSPSC code: 39121600
  • Product type: Spare parts kits
  • Units per package: 1 pk
  • Replacement schedule: As required

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.