Learn More
Grainger Fuse 10A Class D D16 Series Time Delay
Catalog No. 502749636
Encompass_Preferred
The Eaton Bussmann 14L628 is a 14mm x 28mm fast-acting glass tube fuse that provides overload protection for electrical circuits. This time-delay fuse has a voltage rating of 250VAC/DC and a current rating of 6.3A. It features a ceramic body construction and brass endcaps. The 14L628 fuse has a time delay feature that allows harmless overloads to continue for a short time before opening to prevent nuisance tripping. This helps provide protection for motors, power supplies, battery chargers and other electrical equipment against short circuits and overloads. The compact 14x28mm size allows use in tight spaces.
Catalog No. 50-274-9636 Supplier Grainger Supplier No. 14L628
The Eaton Bussmann 14L628 is a 14mm x 28mm fast-acting glass tube fuse that provides overload protection for electrical circuits. This time-delay fuse has a voltage rating of 250VAC/DC and a current rating of 6.3A. It features a ceramic body construction and brass endcaps. The 14L628 fuse has a time delay feature that allows harmless overloads to continue for a short time before opening to prevent nuisance tripping. This helps provide protection for motors, power supplies, battery chargers and other electrical equipment against short circuits and overloads. The compact 14x28mm size allows use in tight spaces.