Learn More
Waters Corp Fuse, 1A, 5 per Package
Catalog No. 50793477 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Waters Corporation fuse is a 1A fuse provided in a package of five. It is designed for compatibility with various Waters instruments, including the 410 Refractive Index Detector, 515 HPLC Pump, and 600 MultiSolvent Delivery System.
  • Features a 1A current rating.
  • Provided in a convenient pack of five.
  • Ensures operational integrity for compatible Waters instruments.
  • Compatible with 410 Refractive Index Detector.
  • Compatible with 515 HPLC Pump.
  • Compatible with 600 MultiSolvent Delivery System.
Catalog No. 50-793-477 Supplier Waters Corp Supplier No. WAT005064
This Waters Corporation fuse is a 1A fuse provided in a package of five. It is designed for compatibility with various Waters instruments, including the 410 Refractive Index Detector, 515 HPLC Pump, and 600 MultiSolvent Delivery System.
  • Features a 1A current rating.
  • Provided in a convenient pack of five.
  • Ensures operational integrity for compatible Waters instruments.
  • Compatible with 410 Refractive Index Detector.
  • Compatible with 515 HPLC Pump.
  • Compatible with 600 MultiSolvent Delivery System.