Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Fuse, 2.0A, 250V, 5 x 20 mm, Fast-Blo

Catalog No. 50793794 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Fuse, 2.0A, 250V, 5 x 20 mm, Fast-Blo

Catalog No. 50-793-794 Supplier Waters Corp Supplier No. WAT16314
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Fuse, 2.0A, 250V, 5 x 20 mm, Fast-Blo

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.