Learn More
Waters Corp Waters Fuse, 3.15 A, 250 V, 5 x 20 mm
Catalog No. 50792631 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This fuse is designed for use in ACQUITY UPLC TUV detectors. It is a 3.15 A, 250 V fuse with dimensions of 5 x 20 mm.
  • Used in ACQUITY UPLC TUV detectors.
  • 3.15 A current rating.
  • 250 V voltage rating.
  • 5 x 20 mm dimensions.
  • Replaces older part numbers used for various detectors and pumps.
  • Available in a 2-pack.
Catalog No. 50-792-631 Supplier Waters Corp Supplier No. 700001800
This fuse is designed for use in ACQUITY UPLC TUV detectors. It is a 3.15 A, 250 V fuse with dimensions of 5 x 20 mm.
  • Used in ACQUITY UPLC TUV detectors.
  • 3.15 A current rating.
  • 250 V voltage rating.
  • 5 x 20 mm dimensions.
  • Replaces older part numbers used for various detectors and pumps.
  • Available in a 2-pack.