Learn More
Grainger Fuse 4A Class D D27 Series Time Delay
Catalog No. 502748659
Encompass_Preferred
The Bussmann IEC Fuse is a 4 amp, 250 volt, fast-acting fuse with a 5x20mm physical size. It has a glass tube construction and features a time-delay design to allow temporary harmless overloads while still providing protection from short circuits and overloads. This fuse has UL and CSA agency approvals and is well-suited for protecting control circuits and other sensitive electronic equipment where fast-acting protection is needed. The compact size allows use in tight spaces.
Catalog No. 50-274-8659 Supplier Grainger Supplier No. 14L630
The Bussmann IEC Fuse is a 4 amp, 250 volt, fast-acting fuse with a 5x20mm physical size. It has a glass tube construction and features a time-delay design to allow temporary harmless overloads while still providing protection from short circuits and overloads. This fuse has UL and CSA agency approvals and is well-suited for protecting control circuits and other sensitive electronic equipment where fast-acting protection is needed. The compact size allows use in tight spaces.