Learn More
Grainger Fuse 63A Class D D33 Series Time Delay
Catalog No. 502752000
Encompass_Preferred
The Bussmann D33 Series 63A Class D Time Delay Fuse is a UL Listed, current-limiting fuse that provides short circuit protection. It has a voltage rating of 600VAC and features a time delay feature that allows temporary harmless overloads while preventing damage from short circuits or severe overloads. This fuse has a 63A amp rating, measures 1-1/4" long x 1/4" diameter, and has 1/4" x 1/4" blade-type end caps. It conforms to UL standard 248-14 and CSA standard C22.2 No. 248.14. The D33 series fuse body is clear to allow for visual inspection of the fusible link.
Catalog No. 50-275-2000 Supplier Grainger Supplier No. 14L626
The Bussmann D33 Series 63A Class D Time Delay Fuse is a UL Listed, current-limiting fuse that provides short circuit protection. It has a voltage rating of 600VAC and features a time delay feature that allows temporary harmless overloads while preventing damage from short circuits or severe overloads. This fuse has a 63A amp rating, measures 1-1/4" long x 1/4" diameter, and has 1/4" x 1/4" blade-type end caps. It conforms to UL standard 248-14 and CSA standard C22.2 No. 248.14. The D33 series fuse body is clear to allow for visual inspection of the fusible link.