Learn More
Grainger Fuse 6A Class D D27 Series Time Delay
Catalog No. 502749717
Encompass_Preferred
The Bussmann Finger-Safe Fuse Block with Cover is a fuse block that provides overcurrent protection for electrical circuits. It has space for up to 6 ATC/ATO automotive blade fuses and features a clear plastic hinged cover for safety. The maximum voltage is 32V DC and maximum current is 40A per circuit. The fuse block has tin-plated brass terminals and measures 2-3/4" L x 1-3/4" W x 1-1/2" H. This finger-safe fuse block is UL recognized and CSA certified. It can be mounted to a surface with screws or attached to a din rail.
Catalog No. 50-274-9717 Supplier Grainger Supplier No. 14L619
The Bussmann Finger-Safe Fuse Block with Cover is a fuse block that provides overcurrent protection for electrical circuits. It has space for up to 6 ATC/ATO automotive blade fuses and features a clear plastic hinged cover for safety. The maximum voltage is 32V DC and maximum current is 40A per circuit. The fuse block has tin-plated brass terminals and measures 2-3/4" L x 1-3/4" W x 1-1/2" H. This finger-safe fuse block is UL recognized and CSA certified. It can be mounted to a surface with screws or attached to a din rail.